Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192006.1 | NP_001178935.1 | 2 - 3 | 392 | 139 | |
XM_008769753.1 | XP_008767975.1 | 3 - 4 | 807 | 139 | ||
XM_008769754.1 | XP_008767976.1 | 4 - 5 | 874 | 139 | ||
XM_008769755.1 | XP_008767977.1 | 3 - 4 | 803 | 139 | ||
XM_008769756.1 | XP_008767978.1 | 3 - 4 | 805 | 139 | ||
XM_008769757.1 | XP_008767979.1 | 2 - 3 | 410 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |