Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192006.1 | NP_001178935.1 | 6 - 7 | 933 | 92 | |
XM_008769753.1 | XP_008767975.1 | 7 - 8 | 1348 | 92 | ||
XM_008769754.1 | XP_008767976.1 | 8 - 9 | 1415 | 92 | ||
XM_008769755.1 | XP_008767977.1 | 7 - 8 | 1326 | 92 | ||
XM_008769756.1 | XP_008767978.1 | 7 - 8 | 1325 | 92 | ||
XM_008769757.1 | XP_008767979.1 | 6 - 7 | 951 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |