Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031708.1 | NP_113896.1 | 4 - 5 | 523 | 92 | |
XM_006235822.1 | XP_006235884.1 | 4 - 5 | 644 | 92 | ||
GenBank mRNA | AB032742.1 | - | 4 - 5 | 523 | 92 | |
BC061773.1 | - | 4 - 5 | 539 | 5621873 | 92 | |
FQ218007.1 | - | 4 - 5 | 541 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |