Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133609.2 | NP_598293.2 | 6 - 7 | 765 | 125 | |
XM_006238630.2 | XP_006238692.1 | 6 - 7 | 736 | 125 | ||
GenBank mRNA | BC072507.1 | - | 6 - 7 | 765 | 7099973 | 125 |
U38253.1 | - | 6 - 7 | 745 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |