Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139465.2 | NP_001132937.1 | 1 - 2 | 239 | 65 | |
GenBank mRNA | CB704217.1 | - | 1 - 2 | 249 | 65 | |
CK476149.1 | - | 1 - 2 | 244 | 65 | ||
FQ212504.1 | - | 1 - 2 | 205 | 65 | ||
FQ214069.1 | - | 1 - 2 | 236 | 65 | ||
FQ231829.1 | - | 1 - 2 | 223 | 65 | ||
FQ232106.1 | - | 1 - 2 | 244 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |