Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008326.1 | NP_001008327.1 | 3 - 4 | 457 | 118 | |
XM_006243001.2 | XP_006243063.1 | 3 - 4 | 371 | 118 | ||
GenBank mRNA | BC086420.1 | - | 3 - 4 | 457 | 7300740 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |