Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015003.2 | NP_001015003.1 | 3 - 4 | 401 | 124 | |
GenBank mRNA | AY973953.1 | - | 3 - 4 | 388 | 124 | |
BC099779.1 | - | 3 - 4 | 386 | 124 | ||
BC127455.1 | - | 3 - 4 | 393 | 7112865 | 124 | |
CB567139.1 | - | 3 - 4 | 411 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |