Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037770.1 | NP_001032859.1 | 2 - 3 | 297 | 61 | |
GenBank mRNA | BC087061.1 | - | 2 - 3 | 327 | 61 | |
BC107670.1 | - | 2 - 3 | 297 | 7937607 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |