Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025754.1 | NP_001020925.1 | 4 - 5 | 736 | 129 | |
XM_006243134.2 | XP_006243196.1 | 4 - 5 | 1550 | 129 | ||
XM_006243135.2 | XP_006243197.1 | 4 - 5 | 1656 | 129 | ||
XM_006243136.2 | XP_006243198.1 | 5 - 6 | 732 | 129 | ||
GenBank mRNA | BC099125.1 | - | 4 - 5 | 736 | 7445605 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |