Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014209.1 | NP_001014231.1 | 1 - 2 | 203 | 132 | |
GenBank mRNA | BC088858.1 | - | 1 - 2 | 203 | 7308076 | 132 |
FQ228964.1 | - | 1 - 2 | 226 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |