Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138863.2 | NP_620218.1 | 3 - 4 | 183 | 75 | |
GenBank mRNA | BC089775.1 | - | 3 - 4 | 183 | 7307365 | 75 |
FQ218841.1 | - | 3 - 4 | 202 | 75 | ||
FQ220898.1 | - | 3 - 4 | 204 | 75 | ||
FQ221589.1 | - | 3 - 4 | 199 | 75 | ||
FQ229147.1 | - | 3 - 4 | 199 | 75 | ||
FQ229679.1 | - | 3 - 4 | 199 | 75 | ||
U66322.2 | - | 2 - 3 | 148 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |