Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025711.1 | NP_001020882.1 | 3 - 4 | 396 | 81 | |
XM_006245533.2 | XP_006245595.1 | 9 - 10 | 936 | 81 | ||
XM_006245534.2 | XP_006245596.1 | 8 - 9 | 839 | 81 | ||
GenBank mRNA | BC098833.1 | - | 3 - 4 | 396 | 7378703 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |