Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017312.2 | NP_059008.1 | 1 - 2 | 66 | 97 | |
GenBank mRNA | BC078871.1 | - | 1 - 2 | 66 | 7132448 | 97 |
HQ696784.1 | - | 1 - 2 | 184 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |