Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245604.1 | XP_006245666.1 | 1 - 2 | 110 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |