Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012728.1 | NP_036860.1 | 7 - 8 | 843 | 94 | |
GenBank mRNA | M97797.1 | - | 7 - 8 | 843 | 94 | |
S75952.1 | - | 7 - 8 | 837 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus 1700 Inventoried Probe spans exons |