Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-Ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008831.3 | NP_001008831.1 | 2 - 3 | 390 | 152 | |
GenBank mRNA | AY626197.1 | - | 2 - 3 | 328 | 152 | |
BC086448.1 | - | 2 - 3 | 340 | 7304077 | 152 | |
FQ225318.1 | - | 2 - 3 | 382 | 152 | ||
FQ225680.1 | - | 2 - 3 | 372 | 152 | ||
FQ225905.1 | - | 2 - 3 | 382 | 152 | ||
FQ226059.1 | - | 2 - 3 | 382 | 152 | ||
FQ226209.1 | - | 2 - 3 | 382 | 152 | ||
FQ226311.1 | - | 2 - 3 | 382 | 152 | ||
FQ227679.1 | - | 2 - 3 | 382 | 152 | ||
FQ227967.1 | - | 2 - 3 | 343 | 152 | ||
FQ228231.1 | - | 2 - 3 | 382 | 152 | ||
FQ229986.1 | - | 2 - 3 | 382 | 152 | ||
FQ231104.1 | - | 2 - 3 | 361 | 152 | ||
FQ231505.1 | - | 2 - 3 | 343 | 152 | ||
FQ231715.1 | - | 2 - 3 | 382 | 152 | ||
FQ231787.1 | - | 2 - 3 | 382 | 152 | ||
FQ231789.1 | - | 2 - 3 | 382 | 152 | ||
FQ232205.1 | - | 2 - 3 | 385 | 152 | ||
FQ232339.1 | - | 2 - 3 | 382 | 152 | ||
FQ232777.1 | - | 2 - 3 | 382 | 152 | ||
FQ232809.1 | - | 2 - 3 | 382 | 152 | ||
FQ233023.1 | - | 2 - 3 | 382 | 152 | ||
FQ233515.1 | - | 2 - 3 | 379 | 152 | ||
FQ234579.1 | - | 2 - 3 | 372 | 152 | ||
FQ234771.1 | - | 2 - 3 | 372 | 152 | ||
KC222945.1 | - | 2 - 3 | 328 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |