Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164744.1 | NP_001158216.1 | 1 - 2 | 64 | 108 | |
GenBank mRNA | DY547565.1 | - | 1 - 2 | 64 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |