Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025750.1 | NP_001020921.1 | 4 - 5 | 677 | 156 | |
XM_006251520.1 | XP_006251582.1 | 4 - 5 | 671 | 156 | ||
GenBank mRNA | BC098846.1 | - | 4 - 5 | 677 | 7379453 | 156 |
FQ225547.1 | - | 4 - 5 | 552 | 156 | ||
FQ232611.1 | - | 4 - 5 | 536 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |