Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173290.1 | NP_775412.1 | 3 - 4 | 896 | 111 | |
GenBank mRNA | AB080317.1 | - | 3 - 4 | 896 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |