Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191599.1 | NP_001178528.1 | 2 - 3 | 457 | 80 | |
XM_006255424.2 | XP_006255486.1 | 2 - 3 | 493 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |