Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251532.2 | XP_006251594.1 | 3 - 4 | 2293 | 104 | |
XM_006251534.2 | XP_006251596.1 | 3 - 4 | 2130 | 104 | ||
XM_008770443.1 | XP_008768665.1 | 4 - 5 | 2447 | 104 | ||
XM_223651.8 | XP_223651.5 | 3 - 4 | 2292 | 104 | ||
GenBank mRNA | FQ230354.1 | - | 2 - 3 | 483 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |