Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037664.1 | NP_001032753.1 | 2 - 3 | 217 | 117 | |
GenBank mRNA | AY623025.1 | - | 1690 | 117 | ||
AY623026.1 | - | 791 | 117 | |||
BC088267.1 | - | 2 - 3 | 217 | 7379090 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |