Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037356.1 | NP_001032433.1 | 2 - 3 | 307 | 109 | |
GenBank mRNA | BC079236.1 | - | 2 - 3 | 287 | 109 | |
BC107922.1 | - | 2 - 3 | 307 | 7444847 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |