Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128183.1 | NP_001121655.1 | 6 - 7 | 542 | 76 | |
GenBank mRNA | DQ537491.1 | - | 6 - 7 | 528 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |