Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012805.2 | NP_036937.2 | 1058 | 84 | ||
GenBank mRNA | AM295012.1 | - | 1058 | 84 | ||
AM295013.1 | - | 1058 | 84 | |||
AM392401.1 | - | 1058 | 84 | |||
L06482.1 | - | 1225 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Rattus norvegicus Probe spans exons |