Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245023.2 | XP_006245085.1 | 2 - 3 | 250 | 80 | |
XM_006245024.2 | XP_006245086.1 | 1 - 2 | 1583 | 80 | ||
XM_006245026.2 | XP_006245088.1 | 2 - 3 | 493 | 80 | ||
XM_006245029.2 | XP_006245091.1 | 2 - 3 | 360 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |