Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100706.1 | NP_001094176.1 | 4 - 5 | 1043 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |