Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106557.1 | NP_001100027.1 | 2 - 3 | 200 | 66 | |
GenBank mRNA | BC158849.1 | - | 2 - 3 | 155 | 7445841 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |