Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ropn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025628.1 | NP_001020799.1 | 4 - 5 | 565 | 90 | |
XM_006248431.2 | XP_006248493.1 | 4 - 5 | 508 | 90 | ||
GenBank mRNA | BC099132.1 | - | 4 - 5 | 565 | 7453834 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |