Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238050.1 | XP_006238112.1 | 5 - 6 | 780 | 73 | |
XM_008763672.1 | XP_008761894.1 | 6 - 7 | 584 | 73 | ||
GenBank mRNA | FQ219084.1 | - | 5 - 6 | 732 | 73 | |
KJ817427.1 | - | 5 - 6 | 598 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |