Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238065.2 | XP_006238127.1 | 1 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |