Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107638.1 | NP_001101108.1 | 6 - 7 | 1057 | 67 | |
XM_006256572.2 | XP_006256634.1 | 6 - 7 | 1354 | 67 | ||
XM_008772989.1 | XP_008771211.1 | 2 - 3 | 1238 | 67 | ||
XM_008772990.1 | XP_008771212.1 | 2 - 3 | 654 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |