Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037656.1 | NP_001032745.1 | 1 - 2 | 236 | 61 | |
GenBank mRNA | BC107937.1 | - | 1 - 2 | 236 | 7320504 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |