Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106576.2 | NP_001100046.2 | 1 - 2 | 280 | 116 | |
GenBank mRNA | BC079464.1 | - | 1 - 2 | 184 | 116 | |
BC161913.1 | - | 1 - 2 | 285 | 8369555 | 116 | |
DY320753.1 | - | 1 - 2 | 280 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |