Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078622.2 | NP_511177.2 | 2 - 3 | 218 | 135 | |
XM_006248444.2 | XP_006248506.1 | 2 - 3 | 371 | 135 | ||
XM_008768694.1 | XP_008766916.1 | 2 - 3 | 569 | 135 | ||
GenBank mRNA | AW919077.1 | - | 2 - 3 | 219 | 135 | |
BC085713.1 | - | 2 - 3 | 216 | 7127059 | 135 | |
L13245.1 | - | 1 - 2 | 115 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |