Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105879.1 | NP_001099349.1 | 9 - 10 | 1285 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |