Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749119.3 | XP_003749167.1 | 6 - 7 | 464 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |