Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109296.1 | NP_001102766.1 | 5 - 6 | 666 | 79 | |
XM_008766561.1 | XP_008764783.1 | 6 - 7 | 1126 | 79 | ||
XM_008766562.1 | XP_008764784.1 | 4 - 5 | 543 | 79 | ||
GenBank mRNA | FQ233502.1 | - | 6 - 7 | 730 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |