Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014096.1 | NP_001014118.1 | 1 - 2 | 254 | 56 | |
GenBank mRNA | BC088427.1 | - | 1 - 2 | 254 | 7124060 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |