Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766445.1 | XP_008764667.1 | 4 - 5 | 384 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |