Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277434.1 | NP_001264363.1 | 6 - 7 | 1314 | 86 | |
GenBank mRNA | FQ227817.1 | - | 4 - 5 | 598 | 86 | |
FQ228265.1 | - | 7 - 8 | 938 | 86 | ||
FQ230804.1 | - | 4 - 5 | 598 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |