Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766577.1 | XP_008764799.1 | 3 - 4 | 516 | 94 | |
XM_576451.6 | XP_576451.4 | 4 - 5 | 593 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |