Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001072998.4 | XP_001072998.2 | 3 - 4 | 492 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |