Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761808.1 | XP_008760030.1 | 8 - 9 | 950 | 94 | |
XM_575125.5 | XP_575125.6 | 8 - 9 | 910 | 94 | ||
GenBank mRNA | FQ213095.1 | - | 8 - 9 | 904 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |