Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022379.1 | NP_071774.1 | 3 - 4 | 503 | 140 | |
XM_006236128.2 | XP_006236190.1 | 2 - 3 | 769 | 140 | ||
GenBank mRNA | L08812.1 | - | 3 - 4 | 503 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |