Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Midn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191577.1 | NP_001178506.1 | 3 - 4 | 387 | 114 | |
XM_006240911.2 | XP_006240973.1 | 4 - 5 | 2044 | 114 | ||
XM_006240912.2 | XP_006240974.1 | 4 - 5 | 1771 | 114 | ||
XM_006240913.2 | XP_006240975.1 | 4 - 5 | 2044 | 114 | ||
XM_008765097.1 | XP_008763319.1 | 3 - 4 | 961 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |