Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230775.2 | XP_006230837.1 | 1 - 2 | 104 | 81 | |
GenBank mRNA | FQ228759.1 | - | 1 - 2 | 74 | 81 | |
FQ231035.1 | - | 1 - 2 | 74 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |