Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759030.1 | XP_008757252.1 | 3 - 4 | 262 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |