Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012624.3 | NP_036756.3 | 8 - 9 | 1193 | 94 | |
XM_006232593.2 | XP_006232655.1 | 8 - 9 | 1324 | 94 | ||
GenBank mRNA | BC161827.1 | - | 7 - 8 | 1183 | 7127842 | 94 |
FQ211172.1 | - | 2 - 3 | 621 | 94 | ||
M11709.1 | - | 8 - 9 | 1194 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |