Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100708.1 | NP_001094178.1 | 10 - 11 | 1468 | 62 | |
XM_006236198.2 | XP_006236260.1 | 10 - 11 | 1516 | 62 | ||
XM_006236200.2 | XP_006236262.1 | 10 - 11 | 1516 | 62 | ||
XM_008762772.1 | XP_008760994.1 | 7 - 8 | 1574 | 62 | ||
XM_008762773.1 | XP_008760995.1 | 7 - 8 | 1573 | 62 | ||
XM_008762774.1 | XP_008760996.1 | 10 - 11 | 1514 | 62 | ||
GenBank mRNA | BC161956.1 | - | 10 - 11 | 1468 | 9027258 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |